Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCF2
- Entrez Gene ID
-
830854
- Gene Name
- ABC transporter family protein
- Gene Aliases
-
ARABIDOPSIS THALIANA GENERAL CONTROL NON-REPRESSIBLE 2, AT5G09930, ATGCN2, ATP-binding cassette F2, GENERAL CONTROL NON-REPRESSIBLE 2, MYH9.14, MYH9_14
- Chromosome Location
-
Chr.5: 3097555 - 3100291 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121030.3 |
NP_196555.2 |
5 - 6 |
1661 |
113 |
View Full Product Details