Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEA6
Entrez Gene ID
831053
Gene Name
K+ efflux antiporter 6
Gene Aliases
ARABIDOPSIS THALIANA K+ EFFLUX ANTIPORTER 6, AT5G11800, ATKEA6, K+ efflux antiporter 6, T22P22.190, T22P22_190
Chromosome Location
Chr.5: 3803315 - 3808273 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121218.5 NP_196741.2 12 - 13 1272 117
GenBank mRNA BT044605.1 - 12 - 13 1068 117

View Full Product Details