Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG32
- Entrez Gene ID
-
817232
- Gene Name
- pleiotropic drug resistance 4
- Gene Aliases
-
AT2G26910, ATP-binding cassette G32, ATPDR4, AtABCG32, F12C20.5, F12C20_5, PDR4, PEC1, PERMEABLE CUTICLE 1, PLEIOTROPIC DRUG RESISTANCE 4, pleiotropic drug resistance 4
- Chromosome Location
-
Chr.2: 11481326 - 11488270 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128248.2 |
NP_180259.1 |
8 - 9 |
1244 |
97 |
View Full Product Details