Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIS4
Entrez Gene ID
817423
Gene Name
histone H4
Gene Aliases
AT2G28740, F8N16.2, F8N16_2, histone H4
Chromosome Location
Chr.2: 12329345 - 12330059 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128434.4 NP_180441.1 1 - 1 512 114
GenBank mRNA AY063857.1 - 1 - 1 448 114

View Full Product Details