Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBL1
Entrez Gene ID
817453
Gene Name
RHOMBOID-like 1
Gene Aliases
AT2G29050, ATRBL1, RHOMBOID-like 1, T9I4.13, T9I4_13
Chromosome Location
Chr.2: 12477966 - 12480374 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128462.4 NP_180469.3 4 - 5 1201 93
GenBank mRNA AK228551.1 - 4 - 5 970 93

View Full Product Details