Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP1
Entrez Gene ID
817489
Gene Name
type one protein phosphatase 1
Gene Aliases
AT2G29400, PP1-AT, PROTEIN PHOSPHATASE 1, type one protein phosphatase 1
Chromosome Location
Chr.2: 12613014 - 12615414 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128494.5 NP_180501.1 1 - 2 360 146
GenBank mRNA AY080871.1 - 1 - 2 335 146
AY142593.1 - 1 - 2 229 146
M93408.1 - 1 - 2 287 146
X64328.1 - - 299 146

View Full Product Details