Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMP1
Entrez Gene ID
821706
Gene Name
transmembrane protein, putative (DUF679 domain membrane protein 1)
Gene Aliases
AT3G21520, Arabidopsis thaliana DUF679 domain membrane protein 1, AtDMP1, DUF679 domain membrane protein 1
Chromosome Location
Chr.3: 7581959 - 7582793 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113047.3 NP_188789.2 1 - 1 650 64
GenBank mRNA AY080599.1 - 1 - 1 650 64
AY114034.1 - 1 - 1 609 64

View Full Product Details