Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TWD1
- Entrez Gene ID
-
821718
- Gene Name
- FKBP-type peptidyl-prolyl cis-trans isomerase family protein
- Gene Aliases
-
AT3G21640, ATFKBP42, FK506-BINDING PROTEIN 42, FKBP42, TWISTED DWARF 1, UCU2, ULTRACURVATA 2
- Chromosome Location
-
Chr.3: 7618851 - 7621477 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113059.3 |
NP_188801.2 |
7 - 8 |
917 |
77 |
GenBank mRNA |
AJ224640.1 |
- |
7 - 8 |
917 |
77 |
View Full Product Details