Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT71B1
Entrez Gene ID
821729
Gene Name
UDP-glucosyl transferase 71B1
Gene Aliases
AT3G21750, UDP-glucosyl transferase 71B1
Chromosome Location
Chr.3: 7664345 - 7666305 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113070.3 NP_188812.1 1 - 1 1748 144
GenBank mRNA AK227147.1 - 1 - 1 1623 144

View Full Product Details