Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKP-ALPHA
Entrez Gene ID
821870
Gene Name
Pyruvate kinase family protein
Gene Aliases
AT3G22960, PKP1, PLASTIDIAL PYRUVATE KINASE 1
Chromosome Location
Chr.3: 8139185 - 8142052 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113196.3 NP_566720.1 1 - 2 787 68
GenBank mRNA AY056793.1 - 1 - 2 711 68
AY058084.1 - 1 - 2 738 68
BT002329.1 - 1 - 2 603 68

View Full Product Details