Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP37
Entrez Gene ID
821886
Gene Name
receptor like protein 37
Gene Aliases
AT3G23110, AtRLP37, EMB2800, EMBRYO DEFECTIVE 2800, receptor like protein 37
Chromosome Location
Chr.3: 8222364 - 8224871 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113212.1 NP_188952.1 1 - 1 1813 93

View Full Product Details