Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VRN5
Entrez Gene ID
822034
Gene Name
Fibronectin type III domain-containing protein
Gene Aliases
AT3G24440, VERNALIZATION 5, VIL1, VIN3-LIKE 1
Chromosome Location
Chr.3: 8875784 - 8878481 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113351.3 NP_189087.1 2 - 3 1025 148
GenBank mRNA EF064791.1 - 2 - 3 715 148

View Full Product Details