Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FKBP15-1
Entrez Gene ID
822115
Gene Name
FK506-binding protein 15 kD-1
Gene Aliases
AT3G25220, FK506-binding protein 15 kD-1
Chromosome Location
Chr.3: 9182616 - 9184679 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113428.3 NP_566762.1 1 - 2 144 92
GenBank mRNA AY062776.1 - 1 - 2 97 92
AY128760.1 - 1 - 2 69 92

View Full Product Details