Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSW3
Entrez Gene ID
836504
Gene Name
Glycosyl hydrolases family 31 protein
Gene Aliases
AT5G63840, MGI19.5, MGI19_5, PRIORITY IN SWEET LIFE 5, PSL5, RADIAL SWELLING 3
Chromosome Location
Chr.5: 25544907 - 25549199 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345613.1 NP_001330435.1 1 - 2 1717 75
NM_125779.3 NP_201189.1 1 - 2 1643 75

View Full Product Details