Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINC2
Entrez Gene ID
837882
Gene Name
nuclear matrix constituent protein-like protein
Gene Aliases
AT1G13220, CROWDED NUCLEI 2, CRWN2, F3F19.25, F3F19_25, LITTLE NUCLEI2
Chromosome Location
Chr.1: 4515464 - 4520306 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332064.1 NP_001322783.1 5 - 6 1342 109
NM_101194.2 NP_563924.1 5 - 6 1107 109

View Full Product Details