Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP3
Entrez Gene ID
838035
Gene Name
purple acid phosphatase 3
Gene Aliases
AT1G14700, ATPAP3, F10B6.10, F10B6_10, purple acid phosphatase 3
Chromosome Location
Chr.1: 5058583 - 5061140 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084069.1 NP_001077538.1 3 - 4 548 75
NM_001332133.1 NP_001320701.1 3 - 4 1069 75
NM_001332134.1 NP_001320700.1 3 - 4 651 75
NM_101339.5 NP_172923.3 3 - 4 651 75
GenBank mRNA AY686599.1 - 3 - 4 554 75
BT000877.1 - 3 - 4 554 75

View Full Product Details