Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBC16
Entrez Gene ID
843880
Gene Name
ubiquitin-conjugating enzyme 16
Gene Aliases
AT1G75440, F1B16.3, F1B16_3, ubiquitin-conjugating enzyme 16
Chromosome Location
Chr.1: 28313379 - 28315122 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106198.5 NP_565110.1 4 - 5 539 73
GenBank mRNA AY065459.1 - 4 - 5 391 73
AY091226.1 - 4 - 5 295 73
DQ027030.1 - 4 - 5 295 73

View Full Product Details