Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PETE1
Entrez Gene ID
843942
Gene Name
plastocyanin 1
Gene Aliases
AT1G76100, T23E18.3, T23E18_3, plastocyanin 1
Chromosome Location
Chr.1: 28553883 - 28554930 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334709.1 NP_001321069.1 1 - 1 845 160
NM_106259.4 NP_177737.1 1 - 1 725 160
GenBank mRNA BT021977.1 - 1 - 1 683 160

View Full Product Details