Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASHH1
Entrez Gene ID
844005
Gene Name
SET domain group 26
Gene Aliases
ASH1 RELATED PROTEIN 1, ASH1-RELATED PROTEIN 1, AT1G76710, F28O16.8, F28O16_8, SDG26, SET domain group 26
Chromosome Location
Chr.1: 28789698 - 28792719 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106321.4 NP_177797.2 10 - 11 1337 66
NM_202429.2 NP_974158.1 9 - 10 1466 66
GenBank mRNA AB493538.1 - 9 - 10 1197 66
BT001913.1 - 9 - 10 1197 66

View Full Product Details