Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD3
Entrez Gene ID
844201
Gene Name
DNA-directed DNA polymerase
Gene Aliases
AT1G78650, T30F21.2, T30F21_2
Chromosome Location
Chr.1: 29582907 - 29585665 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106512.3 NP_565185.1 7 - 8 1468 62

View Full Product Details