Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP79C1
- Entrez Gene ID
-
844275
- Gene Name
- cytochrome P450, family 79, subfamily C, polypeptide 1
- Gene Aliases
-
''cytochrome P450, AT1G79370, YUP8H12R.1, YUP8H12R_1, cytochrome P450, family 79, polypeptide 1, polypeptide 1'', subfamily C
- Chromosome Location
-
Chr.1: 29857934 - 29860163 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106585.2 |
NP_178055.2 |
2 - 3 |
995 |
92 |
View Full Product Details