Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX2
Entrez Gene ID
844278
Gene Name
cation/H+ exchanger 2
Gene Aliases
AT1G79400, ATCHX2, T8K14.18, T8K14_18, cation/H+ exchanger 2
Chromosome Location
Chr.1: 29864992 - 29867840 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106588.1 NP_178058.1 2 - 3 1146 89

View Full Product Details