Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT3
Entrez Gene ID
844308
Gene Name
nudix hydrolase homolog 3
Gene Aliases
AT1G79690, F20B17.11, F20B17_11, atnudt3, nudix hydrolase homolog 3
Chromosome Location
Chr.1: 29985046 - 29990388 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334912.1 NP_001320544.1 19 - 20 2164 88
NM_106618.5 NP_565218.1 18 - 19 2336 88
GenBank mRNA AY120774.1 - 18 - 19 2052 88
BT000142.1 - 18 - 19 2022 88

View Full Product Details