Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT3
Entrez Gene ID
844308
Gene Name
nudix hydrolase homolog 3
Gene Aliases
AT1G79690, F20B17.11, F20B17_11, atnudt3, nudix hydrolase homolog 3
Chromosome Location
Chr.1: 29985046 - 29990388 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334912.1 NP_001320544.1 2 - 3 207 75
NM_106618.5 NP_565218.1 1 - 2 379 75
GenBank mRNA AY120774.1 - 1 - 2 95 75
BT000142.1 - 1 - 2 65 75

View Full Product Details