Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DML2
Entrez Gene ID
820162
Gene Name
demeter-like 2
Gene Aliases
AT3G10010, demeter-like 2
Chromosome Location
Chr.3: 3081735 - 3088550 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337853.1 NP_001326647.1 6 - 7 1880 65
NM_111836.5 NP_187612.5 5 - 6 1750 65

View Full Product Details