Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DFC
- Entrez Gene ID
-
820179
- Gene Name
- DHFS-FPGS homolog C
- Gene Aliases
-
A. THALIANA DHFS-FPGS HOMOLOG C, AT3G10160, ATDFC, DHFS-FPGS homolog C, FPGS2, folylpolyglutamate synthetase 2
- Chromosome Location
-
Chr.3: 3139383 - 3144615 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111851.6 |
NP_187627.3 |
7 - 8 |
1031 |
154 |
View Full Product Details