Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT3G10780
Entrez Gene ID
820247
Gene Name
emp24/gp25L/p24 family/GOLD family protein
Gene Aliases
AT3G10780
Chromosome Location
Chr.3: 3375105 - 3376335 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111915.3 NP_187689.2 3 - 4 564 81

View Full Product Details