Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSD1
Entrez Gene ID
820263
Gene Name
manganese superoxide dismutase 1
Gene Aliases
ARABIDOPSIS MANGANESE SUPEROXIDE DISMUTASE 1, AT3G10920, ATMSD1, AtSOD1, MANGANESE SUPEROXIDE DISMUTASE 1, MATERNAL EFFECT EMBRYO ARREST 33, MEE33, manganese superoxide dismutase 1, superoxide dismutase 1
Chromosome Location
Chr.3: 3417940 - 3419892 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111929.4 NP_187703.1 3 - 4 541 91
GenBank mRNA AY059807.1 - 3 - 4 493 91
AY072495.1 - 3 - 4 466 91

View Full Product Details