Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATE2
Entrez Gene ID
820295
Gene Name
arginine-tRNA protein transferase 2
Gene Aliases
AT3G11240, ATATE2, arginine-tRNA protein transferase 2
Chromosome Location
Chr.3: 3518472 - 3521188 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337921.1 NP_001326208.1 1 - 2 240 106
NM_111959.3 NP_187733.1 1 - 2 240 106
GenBank mRNA AF370200.1 - 1 - 2 167 106
AY056368.1 - 1 - 2 144 106

View Full Product Details