Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYB21
Entrez Gene ID
822401
Gene Name
myb domain protein 21
Gene Aliases
ARABIDOPSIS THALIANA MYB DOMAIN PROTEIN 3, ARABIDOPSIS THALIANA MYB DOMAIN PROTEIN 3, AT3G27810, AT3G27812, ATMYB21, ATMYB3, myb domain protein 21
Chromosome Location
Chr.3: 10305849 - 10310492 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338911.1 NP_001326661.1 1 - 2 399 88
NM_113696.3 NP_189418.2 2 - 3 2033 88
GenBank mRNA AB493635.1 - 2 - 3 286 88
AK118439.1 - 2 - 3 420 88
AY519589.1 - 2 - 3 286 88

View Full Product Details