Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQR
Entrez Gene ID
822411
Gene Name
NADPH:quinone oxidoreductase
Gene Aliases
AT3G27890, NADPH:QUINONE OXIDOREDUCTASE, NADPH:quinone oxidoreductase
Chromosome Location
Chr.3: 10350595 - 10352033 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113705.4 NP_189427.1 1 - 2 195 74
GenBank mRNA BT024581.1 - 1 - 2 100 74

View Full Product Details