Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCA7
- Entrez Gene ID
-
823932
- Gene Name
- ABC2 homolog 6
- Gene Aliases
-
A. THALIANA ABC2 HOMOLOG 6, ABC2 homolog 6, AT3G47780, ATATH6, ATH6, ATP-binding cassette A7
- Chromosome Location
-
Chr.3: 17624340 - 17629285 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114646.4 |
NP_190362.2 |
9 - 10 |
1520 |
90 |
View Full Product Details