Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLDH
Entrez Gene ID
823948
Gene Name
L-galactono-1,4-lactone dehydrogenase
Gene Aliases
''L-galactono-1, 4-LACTONE DEHYDOROGENASE, 4-lactone dehydrogenase, 4-lactone dehydrogenase'', AT3G47930, ATGLDH, L-GALACTONO-1, L-galactono-1
Chromosome Location
Chr.3: 17684322 - 17687661 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339340.1 NP_001319706.1 3 - 4 1237 75
NM_114662.3 NP_190376.1 3 - 4 1288 75

View Full Product Details