Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH2B4
Entrez Gene ID
823955
Gene Name
aldehyde dehydrogenase 2B4
Gene Aliases
ALDEHYDE DEHYDROGENASE 2, ALDH2, ALDH2A, AT3G48000, aldehyde dehydrogenase 2, aldehyde dehydrogenase 2A, aldehyde dehydrogenase 2B4
Chromosome Location
Chr.3: 17716864 - 17720169 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114669.4 NP_190383.1 6 - 7 1156 62
GenBank mRNA AB030820.1 - 6 - 7 912 62
AF327426.1 - 6 - 7 906 62
AF349447.1 - 6 - 7 912 62
AF349522.1 - 6 - 7 830 62
AF372911.1 - 6 - 7 906 62
AY090443.1 - 6 - 7 910 62
BT000752.1 - 6 - 7 855 62
BT006371.1 - 6 - 7 830 62

View Full Product Details