Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAT3
Entrez Gene ID
824130
Gene Name
branched-chain aminotransferase 3
Gene Aliases
AT3G49680, ATBCAT-3, branched-chain aminotransferase 3
Chromosome Location
Chr.3: 18422542 - 18425758 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125330.1 NP_001118802.1 2 - 3 320 67
NM_114828.6 NP_566923.1 2 - 3 433 67
GenBank mRNA AF446355.1 - 2 - 3 298 67
AJ276124.1 - 2 - 3 300 67
AY097417.1 - 2 - 3 207 67

View Full Product Details