Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GH9C3
Entrez Gene ID
826706
Gene Name
glycosyl hydrolase 9C3
Gene Aliases
AT4G11050, AtGH9C3, T22B4.30, T22B4_30, glycosyl hydrolase 9C3
Chromosome Location
Chr.4: 6747234 - 6751536 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340700.1 NP_001328090.1 6 - 7 1383 76
NM_001340701.1 NP_001328091.1 6 - 7 1372 76
NM_117175.3 NP_192843.2 6 - 7 1383 76
GenBank mRNA AY133685.1 - 6 - 7 1158 76

View Full Product Details