Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9C3
Entrez Gene ID
826706
Gene Name
glycosyl hydrolase 9C3
Gene Aliases
AT4G11050, AtGH9C3, T22B4.30, T22B4_30, glycosyl hydrolase 9C3
Chromosome Location
Chr.4: 6747234 - 6751536 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340700.1 NP_001328090.1 7 - 8 1734 109
NM_001340701.1 NP_001328091.1 7 - 8 1723 109
NM_117175.3 NP_192843.2 7 - 8 1734 109
GenBank mRNA AY133685.1 - 7 - 8 1509 109

View Full Product Details