Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FEY
Entrez Gene ID
828890
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT4G27760, FEY3, FOREVER YOUNG
Chromosome Location
Chr.4: 13844059 - 13846746 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118915.6 NP_194506.4 6 - 7 891 70

View Full Product Details