Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP1
Entrez Gene ID
832157
Gene Name
Ankyrin repeat family protein with DHHC zinc finger domain-containing protein
Gene Aliases
AT5G20350, F5O24.240, F5O24_240, TIP GROWTH DEFECTIVE 1
Chromosome Location
Chr.5: 6876441 - 6881506 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122042.4 NP_197535.2 3 - 4 846 71
GenBank mRNA AK228596.1 - 3 - 4 698 71
AY965346.1 - 3 - 4 515 71
JQ937239.1 - 3 - 4 515 71

View Full Product Details