Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRXL4
Entrez Gene ID
832176
Gene Name
BREVIS RADIX-like 4
Gene Aliases
AT5G20540, ATBRXL4, BREVIS RADIX-like 4, BRX-LIKE4, F7C8.130, F7C8_130
Chromosome Location
Chr.5: 6947789 - 6952038 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122061.4 NP_197554.2 1 - 2 250 89
GenBank mRNA AK117133.1 - 1 - 2 196 89

View Full Product Details