Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TSL
- Entrez Gene ID
-
832217
- Gene Name
- Protein kinase superfamily protein
- Gene Aliases
-
AT5G20930, F22D1.100, F22D1_100, PROTEIN KINASE TSL, TOUSLED
- Chromosome Location
-
Chr.5: 7097868 - 7103206 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122101.2 |
NP_568405.1 |
8 - 9 |
1272 |
70 |
View Full Product Details