Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET12
Entrez Gene ID
832367
Gene Name
tetraspanin12
Gene Aliases
AT5G23030, MYJ24.2, MYJ24_2, tetraspanin12
Chromosome Location
Chr.5: 7726702 - 7728369 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122209.4 NP_197694.1 1 - 2 613 93
GenBank mRNA BT010941.1 - 1 - 2 527 93
BT011655.1 - 1 - 2 496 93

View Full Product Details