Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-B12
Entrez Gene ID
832527
Gene Name
phloem protein 2-B12
Gene Aliases
AT5G24560, AtPP2-B12, K18P6.9, K18P6_9, phloem protein 2-B12
Chromosome Location
Chr.5: 8402205 - 8403486 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343880.1 NP_001318638.1 2 - 3 369 70

View Full Product Details