Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRY3
Entrez Gene ID
832554
Gene Name
cryptochrome 3
Gene Aliases
AT5G24850, F6A4.60, F6A4_60, cryptochrome 3
Chromosome Location
Chr.5: 8535258 - 8538211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122394.4 NP_568461.3 2 - 3 606 169

View Full Product Details