Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SK 11
Entrez Gene ID
832733
Gene Name
shaggy-related kinase 11
Gene Aliases
ARABIDOPSIS THALIANA SHAGGY-RELATED KINASE 11, ASKALPHA, ASKalpha, AT5G26751, ATSK11, Arabidopsis GSK alpha, F2P16.21, F2P16_21, shaggy-related kinase 11
Chromosome Location
Chr.5: 9399377 - 9402476 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122557.3 NP_568486.1 4 - 5 499 101
GenBank mRNA AF428327.1 - 4 - 5 499 101
AY046024.1 - 4 - 5 499 101
AY142595.1 - 3 - 4 256 101

View Full Product Details