Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX6B
Entrez Gene ID
838851
Gene Name
cytochrome C oxidase 6B
Gene Aliases
AT1G22450, ATCOX6B2, CYTOCHROME C OXIDASE 6B2, F12K8.20, F12K8_20, SUBUNIT 6B OF CYTOCHROME C OXIDASE, cytochrome C oxidase 6B
Chromosome Location
Chr.1: 7925121 - 7927345 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102094.4 NP_173661.1 2 - 3 682 83
GenBank mRNA AB035444.1 - 2 - 3 384 83
AY063997.1 - 2 - 3 489 83
AY096696.1 - 2 - 3 356 83

View Full Product Details