The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332597.1 | NP_001320610.1 | 4 - 5 | 722 | 105 |
NM_102175.4 | NP_564191.1 | 4 - 5 | 573 | 105 | |
GenBank mRNA | AY059896.1 | - | 4 - 5 | 535 | 105 |
AY093340.1 | - | 4 - 5 | 421 | 105 |