Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO14
Entrez Gene ID
839211
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT1G26700, ATMLO14, MILDEW RESISTANCE LOCUS O 14, T24P13.8, T24P13_8
Chromosome Location
Chr.1: 9227378 - 9232437 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332726.1 NP_001319086.1 6 - 7 979 109
NM_102433.1 NP_564257.1 5 - 6 670 109
GenBank mRNA AF369575.1 - 5 - 6 670 109

View Full Product Details