Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ORC6
- Entrez Gene ID
-
839227
- Gene Name
- origin recognition complex protein 6
- Gene Aliases
-
ARABIDOPSIS THALIANA ORIGIN RECOGNITION COMPLEX PROTEIN 6, AT1G26840, ATORC6, ORIGIN RECOGNITION COMPLEX SUBUNIT 6, T2P11.3, T2P11_3, origin recognition complex protein 6
- Chromosome Location
-
Chr.1: 9298670 - 9300800 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102448.4 |
NP_174006.2 |
4 - 5 |
505 |
71 |
GenBank mRNA |
AJ575584.1 |
- |
4 - 5 |
353 |
71 |
View Full Product Details