Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIM5
Entrez Gene ID
833544
Gene Name
fimbrin-like protein 2
Gene Aliases
AT5G35700, FIM2, FIMBRIN5, MXH1.5, MXH1_5, fimbrin-like protein 2
Chromosome Location
Chr.5: 13872479 - 13876876 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122961.4 NP_198420.1 14 - 15 2034 89
GenBank mRNA AY045780.1 - 14 - 15 2012 89
AY142570.1 - 13 - 14 1809 89

View Full Product Details